5-Carbethoxyuracil

Lieferant: Apollo Scientific

APOSOR322325-1GEA 56.8 EUR
APOSOR322325-1G
5-Carbethoxyuracil
5-Carbethoxyuracil
Formel: C₇H₈N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00057337
CAS-Nummer: 28485-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR