Ethylhippurat, technische Qualität

Lieferant: Apollo Scientific

APOSOR32420-1GEA 51 EUR
APOSOR32420-1G
Ethylhippurat, technische Qualität
Ethylhippurat
Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026890
CAS-Nummer: 1499-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR