L-Glutaminsäure-5-methylester

Lieferant: Apollo Scientific

APOSOR350212-100GEA 77.1 EUR
APOSOR350212-100G
L-Glutaminsäure-5-methylester
L-Glutaminsäure-5-methylester
Formel: C₆H₁₁NO₄
Molecular Weight: 161,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002632
CAS-Nummer: 1499-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR