Tris(4-nitrophenyl)amine

Lieferant: Apollo Scientific

APOSOR350395-100GEA 292 EUR
APOSOR350395-100G
Tris(4-nitrophenyl)amine
Tris(4-nitrophenyl)amine
Formel: C₁₈H₁₂N₄O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00024636
CAS-Nummer: 20440-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR