4-Ethinylanilin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Aminophenyl)acetylen, 1-Ethinyl-4-aminobenzol, (4-Aminophenyl)ethin, 1-Amino-4-ethinylbenzol

OR400295-25G
APOSOR400295-25GEA 106 EUR
APOSOR400295-25G
4-Ethinylanilin 98%
4-Ethinylanilin
Formel: C₈H₇N
Molecular Weight: 117,15 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00168824
CAS-Nummer: 14235-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR