4-Brom-3-methoxybenzonitril
Lieferant: Apollo Scientific
Synonyme:
4-Brom-3-methoxybenzoesäurenitril
APOSOR46048-10GEA
111
EUR
APOSOR46048-10G
APOSOR46048-1G
APOSOR46048-5G
4-Brom-3-methoxybenzonitril
4-Brom-3-methoxybenzonitril
Formel:
C₈H₆BrNO Molecular Weight: 212,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09753723 CAS-Nummer: 120315-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...