Tetraethylammoniumbromid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetrylammoniumbromid

APOSOR4671-1KGEA 109 EUR
APOSOR4671-1KG APOSOR4671-500G
Tetraethylammoniumbromid 97%
Tetraethylammoniumbromid
Formel: C₈H₂₀BrN
Molecular Weight: 210,16 g/mol
Schmelzpunkt: 285…287 °C
Dichte: 1,4 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011825
CAS-Nummer: 71-91-0
EINECS: 200-769-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR