N-Fmoc-glycin 98%
Lieferant: Apollo Scientific
APOSOR54569-500GEA
219
EUR
APOSOR54569-500G
APOSOR54569-100G
N-Fmoc-glycin 98%
N-Fmoc-glycin
Formel:
C₁₇H₁₅NO₄ Molecular Weight: 297,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037140 CAS-Nummer: 29022-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...