3-(2-Chlorphenyl)-5-isoxazolcarbonsäure

Lieferant: Apollo Scientific

APOSOR60173-250MGEA 364 EUR
APOSOR60173-250MG
3-(2-Chlorphenyl)-5-isoxazolcarbonsäure
3-(2-Chlorophenyl)-5-isoxazolecarboxylic acid
Formel: C₁₀H₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01935954
CAS-Nummer: 338982-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR