1,3-Benzoxazol-6-amin

Lieferant: Apollo Scientific
Achtung

APOSOR70136-1GEA 77.1 EUR
APOSOR70136-1G
1,3-Benzoxazol-6-amin
1,3-Benzoxazol-6-amin
Formel: C₇H₆N₂O
Molecular Weight: 134,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03791187
CAS-Nummer: 177492-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR