8-Brom-2-chlorchinolin 97%

Lieferant: Apollo Scientific
Achtung

APOSOR8709-1GEA 81.5 EUR
APOSOR8709-1G
8-Brom-2-chlorchinolin 97%
8-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,5000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08741410
CAS-Nummer: 163485-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR