N-Benzylacetamide 98%

Lieferant: Apollo Scientific

Synonyme: Lacosamide EP Impurity G, N-Acetylbenzylamine

OR904376-100G
APOSOR904376-100GEA 186 EUR
APOSOR904376-100G
N-Benzylacetamide 98%
N-Benzylacetamide
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059204
CAS-Nummer: 588-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR