Ethyl(trimethylsilyl)acetat 95%

Lieferant: Apollo Scientific

Synonyme: (Trimethylsilyl)essigsäureethylester

OR921875-100G
APOSOR921875-100GEA 88.6 EUR
APOSOR921875-100G
Ethyl(trimethylsilyl)acetat 95%
Ethyl(trimethylsilyl)acetat
Formel: C₇H₁₆O₂Si
Molecular Weight: 160,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009172
CAS-Nummer: 4071-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR