1-Phenyl-1-(trimethylsilyloxy)ethylen 95%

Lieferant: Apollo Scientific

APOSOR928812-1GEA 132 EUR
APOSOR928812-1G APOSOR928812-5G APOSOR928812-10G
1-Phenyl-1-(trimethylsilyloxy)ethylen 95%
1-Phenyl-1-(trimethylsilyloxy)ethylen
Formel: C₁₁H₁₆OSi
Molecular Weight: 192,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008582
CAS-Nummer: 13735-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR