5-Iodisatin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Jodindolin-2,3-dion
, 5-Jod-1H-indol-2,3-dion
, 5-Iodindolin-2,3-dion
, 5-Iod-1H-indol-2,3-dion
, 5-Jod-2,3-indolindion
, 5-Jod-2,3-dioxoindolin
, 5-Iod-2,3-dioxoindolin
, 5-Iod-2,3-indolindion
APOSOR9765-5GEA
17.6
EUR
APOSOR9765-5G
APOSOR9765-100G
APOSOR9765-25G
5-Iodisatin 97%
5-Iodisatin
Formel:
C₈H₄INO₂ Molecular Weight: 273,03 g/mol Schmelzpunkt: 276…280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016899 CAS-Nummer: 20780-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...