3-Hydrazinobenzoesäure

Lieferant: Apollo Scientific
Achtung

APOSOR9870-25GEA 97.4 EUR
APOSOR9870-25G APOSOR9870-10G APOSOR9870-50G
3-Hydrazinobenzoesäure
3-Hydrazinobenzoic acid
Formel: C₇H₈N₂O₂
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 191…194 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00191447
CAS-Nummer: 38235-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR