3-Nitro-L-tyrosin 98%
Lieferant: Apollo Scientific
APOSOR9954-25GEA
78.6
EUR
APOSOR9954-25G
3-Nitro-L-tyrosin 98%
3-Nitro-L-tyrosin
Formel:
C₉H₁₀N₂O₅ Molecular Weight: 226,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007123 CAS-Nummer: 621-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...