1-(3-(Trifluormethyl)phenyl)thioharnstoff 97%

Lieferant: Apollo Scientific
Achtung

APOSPC0942-25GEA 147 EUR
APOSPC0942-25G
1-(3-(Trifluormethyl)phenyl)thioharnstoff 97%
1-(3-(Trifluormethyl)phenyl)thioharnstoff
Formel: C₈H₇F₃N₂S
Molecular Weight: 220,22 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041171
CAS-Nummer: 1736-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR