2-Chlor-5-fluoranisol 97%
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC1824E-10GEA
97.4
EUR
APOSPC1824E-10G
2-Chlor-5-fluoranisol 97%
2-Chlor-5-fluoranisol
Formel:
C₇H₆ClFO Molecular Weight: 160,58 g/mol Siedepunkt: 195 °C Dichte: 1,3 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042573 CAS-Nummer: 450-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...