Kaliumethyltrifluoroborat

Lieferant: Apollo Scientific

APOSPC200381-1GEA 75.7 EUR
APOSPC200381-1G
Kaliumethyltrifluoroborat
Potassium ethyltrifluoroborate
Formel: C₂H₅BF₃K
Molecular Weight: 135,9656 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112713
CAS-Nummer: 44248-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR