(3-Chlor-5-fluor-4-methoxyphenyl)essigsäure 98%

Lieferant: Apollo Scientific

PC302718-1G PC302718-5G
APOSPC302718-1GEA 180 EUR
APOSPC302718-1G APOSPC302718-5G
(3-Chlor-5-fluor-4-methoxyphenyl)essigsäure 98%
(3-Chlor-5-fluor-4-methoxyphenyl)essigsäure
Formel: C₉H₈ClFO₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1000544-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR