3,5-Dinitro-4-fluorbenzotrifluorid 98%

Lieferant: Apollo Scientific

APOSPC3040H-1GEA 306 EUR
APOSPC3040H-1G APOSPC3040H-5G
3,5-Dinitro-4-fluorbenzotrifluorid 98%
3,5-Dinitro-4-fluorbenzotrifluorid
Formel: C₇H₃F₃N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00153131
CAS-Nummer: 393-76-0
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR