1-(2-Bromethoxy)-3-(trifluormethyl)benzol

Lieferant: Apollo Scientific
Achtung

APOSPC4638-1GEA 43.7 EUR
APOSPC4638-1G
1-(2-Bromethoxy)-3-(trifluormethyl)benzol
1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene
Formel: C₉H₈BrF₃O
Molecular Weight: 269,06 g/mol
Siedepunkt: 116…118 °C/10mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00215501
CAS-Nummer: 18800-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR