4-Toluolsulfonyl-(3-trifluormethylanilid)

Lieferant: Apollo Scientific

APOSPC6301-1GEA 256 EUR
APOSPC6301-1G
4-Toluolsulfonyl-(3-trifluormethylanilid)
4-Toluolsulfonyl-(3-trifluormethylanilid)
Formel: C₁₄H₁₂F₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00245086
CAS-Nummer: 1584-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR