Copper(II) gluconate ≥98%, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL344419-100GEA 83.8 EUR
SIAL344419-100G
Copper(II) gluconate ≥98%, Sigma-Aldrich®
Copper(II) gluconate

Copper(II) D-gluconate powder in combination with silicon was used for the fabrication of carbon coated Si-Cu3Si composite as the anode materials for Li-ion batteries.

Formel: C₁₂H₂₂CuO₁₄
Molecular Weight: 453,84 g/mol
Schmelzpunkt: 155-157 °C (Lit.)
MDL: MFCD00075297
CAS-Nummer: 527-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR