DNA (Deoxyribonucleic acid) (aus Lachssperma)

Lieferant: STRATAGENE EUROPE

STRA201190EA 479 EUR
STRA201190
DNA (Deoxyribonucleic acid) (aus Lachssperma)
DNS (Desoxyribonukleinsäure)

Kit contains salmon sperm DNA in its liquid form.


  • Have pH of 7,5
  • Melting point: 0 °C
  • Boiling point: 100 °C


It can easily soluble in cold water and hot water.

MDL: MFCD00130922
CAS-Nummer: 9007-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR