Blood sedimentation apparatus, stainless steel, MACRO, Assistent
Lieferant: Hecht
Assistent®
HECH40367005EA 112.7 EUR
HECH40367005
Blood sedimentation apparatus, stainless steel, MACRO, Assistent
Pipetten Haemometerpipetten
With tilting device for rapid sedimentation.
Complete with blood sedimentation pipettes, protecting caps for pipettes, blood mixing tubes with rubber stoppers and special rubber caps.