Petroleumbenzin, 50…70 °C ≥90%, AnalaR NORMAPUR®

Lieferant: VWR Chemicals
Gefahr

Synonyme: BenzinPetrolether

88263.290EA 62.7 EUR
88263.290 88263.460 88263.360
Petroleumbenzin, 50…70 °C ≥90%, AnalaR NORMAPUR®
Petroleumbenzin
MDL: MFCD00081849
CAS-Nummer: 64742-49-0
UN: 1268
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Readily carbonisable substances passes test
Density (20/4) 0.654 - 0.670
Free acidity max. 0.00008 meq/g
Boiling range (50-70°C) (Vol%) min. 90%
Evaporation residue max. 10 ppm
Water max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR