PTFE tubing, BOLA
Lieferant: Bohlender
BOLA
BOHLS1810-41EA 29.4 EUR
BOHLS1810-41
PTFE tubing, BOLA
Schläuche Transferschlauch
Translucent to milky-white appearance.
  • Almost universal chemical resistance
  • Free from extractable agents
  • Physiologically safe
  • Flame retardant according to UL94V0
Perfect tubing for aggressive and pure liquids or gases.