Economical Carbon-Impregnated Paper Cartridge
Lieferant: Avantor Fluid Handling
01509-25 01509-27
MFLX01509-25EA 20.1 EUR
MFLX01509-25 MFLX01509-27
Economical Carbon-Impregnated Paper Cartridge
Filter Filterkartuschen
Ideal for chemically neutral waters with lower chlorine concentrations.
  • Constructed of carbon-impregnated cellulose
This cellulose cartridge removes organic tastes/odors and chlorine, plus reduces sediment. Suitable to 125 °F (52 °C).