High-Flow Polypropylene Felt Filter Bags
Lieferant: Pentair
MFLX01523-46EA 6.4 EUR
MFLX01523-46
High-Flow Polypropylene Felt Filter Bags
Filter Bag Filter Systems
Wide range of options for temperature, flow rate, and filtration quality.
  • Constructed of Polypropylene Felt
Designed for higher flow with an average efficiency at the rated pore size. Polypropylene felt is ideal for general-purpose filtration. The bags are glazed to minimize fiber migration. Polypropylene felt filter bags are rated for 200 °F (93 °C) maximum.