Parker Hannifin Push-to-Connect Check Valves, Acetal
Lieferant: Parker Hannifin
MFLX01379-89EA 21.6 EUR
MFLX01379-89 MFLX01379-93 MFLX01379-91
Parker Hannifin Push-to-Connect Check Valves, Acetal
Ventile Rückschlagventile
Create a secure connection with your check valve.
  • Constructed of durable acetal plastic
Use these push-to-connect check valves for rigid and semirigid plastic tubing. Acetal body with EPDM O-rings. Temperature range of 34 °F (1.1 °C) to 150 °F (65.6 °C).