Walchem High Stroke-Frequency Diaphragm Metering Pump Service Kits
Lieferant: Walchem
MFLX74125-18EA 151.44 EUR
MFLX74125-18 MFLX74125-08 MFLX74126-18 MFLX74125-38 MFLX74126-28 MFLX74125-28 MFLX74126-38
Walchem High Stroke-Frequency Diaphragm Metering Pump Service Kits
Pumpen Membranpumpen
Extend the life of your pump.
  • Easy and quick maintenance
Easy to install, these service kits avoid costly downtime of your pump.
Lieferumfang: Includes diaphragm and retainer, valve guides, valve seats, valve balls, valve gaskets, and o-rings.