2,2',3,4,4',5,6,6'-Octachlorbiphenyl (PCB Nr. 204)

Lieferant: LGC Standards PROMOCHEM

PROOCIL-PCB-204-CEA 652 EUR
PROOCIL-PCB-204-C
2,2',3,4,4',5,6,6'-Octachlorbiphenyl (PCB Nr. 204)
2,2',3,4,4',5,6,6'-Octachlorbiphenyl (PCB Nr. 204)
Formel: C₁₂H₂Cl₈
Molecular Weight: 429,77 g/mol
CAS-Nummer: 74472-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR