TapeStation Parts and Accessories for 2200
Lieferant: Agilent Technologies
AGLS5067-5154EA 235 EUR
AGLS5067-5154 AGLS5067-5153 AGLS5067-5152 AGLS5067-5150 AGLS5067-5601 AGLS5067-5155
TapeStation Parts and Accessories for 2200
Elektrophorese-Systeme Kapillarelektrophorese-Systeme
Accessories and spare parts for the 2200 TapeStation system like pates and foil seals, loading tips, TapeStation test tape, needle cartridge.
  • 8-well PCR tube strips or 96-well sample plates are available depending on sample throughput, bringing added flexibility
  • Needle cartridges enable fast and simple needle exchange to ensure proper maintenance of the 2200 TapeStation system
  • TapeStation test tape is available for troubleshooting and running system diagnostics tests. The Tapes are reusable and expire after two years
Our portfolio of 2200 TapeStation spare parts and accessories features everything you need to keep your lab productive. Maximise the efficiency of your 2200 TapeStation system today with our genuine agilent accessories.
The portfolio features additional needle cartridges and TapeStation test tape. In addition, if running short on spare parts, extra loading tips are also available, along with 96-well sample plates and foil seals. For enhanced flexibility we also offer 8-well PCR optical tube strips and caps.
Please note that the support for agilent 2200 TapeStation systems ends on december 31, 2022. For more information, please contact your local agilent representative.
Bitte beachten For research use only. Not for use in diagnostic procedures.