DermaLife™ K Keratinocyte Medium Kit
Lieferant: Lifeline Cell Technology
CELSLL-0007EA 187 EUR
CELSLL-0007
DermaLife™ K Keratinocyte Medium Kit
Zellkulturmedien
DermaLife™ K has been optimised for the culture of Human Epidermal Keratinocytes in a serum-free environment.
  • Contains supplements and growth factors
  • Grows human epidermal keratinocytes
  • Maintains excellent cell morphology
  • Contains no phenol red or antimicrobials
DermaLife™ K LifeFactors® Kit contains supplements and growth factors for use with human keratinocytes and DermaLife basal medium. DermaLife™ K Medium grows human epidermal keratinocytes through at least 15 population doublings at rates that meet or exceed other commercially available serum-free media while maintaining excellent cell morphology. DermaLife™ K contains no phenol red or antimicrobials.
Lifeline® optimised culture media contain no antimicrobials and no phenol red.
Bestellinformation: DermaLife™ K Medium Complete Kit includes DermaLife basal medium and DermaLife K LifeFactors® Kit of associated supplements and growth factors. The LifeFactors® include L-Glutamine 15 ml, Extract P® 2 ml, Epinephrine 0,5 ml, rh TGF-α 0,5 ml, Hydrocortisone Hemisuccinate 0,5 ml, rh Insulin 0,5 ml, Apo-Transferrin 0,5 ml, Calcium Chloride 0,5 ml, Gentamicin and Amphotericin B 0,5 ml.