Potassium sorbate ≥98,0%, Pulver FCC

Lieferant: Spectrum Chemical

Synonyme: trans,trans-2,4-Hexadiensäure KaliumsalzKalium SorbicumKalium-(E,E)-2,4-hexadienoatSorbinsäure Kaliumsalz

SPCMP1408-125GMEA 128 EUR
SPCMP1408-125GM
Potassium sorbate ≥98,0%, Pulver FCC
Kaliumsorbat

Potassium Sorbate, Powder, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formel: CH₃CH=CHCH=CHCOOK
Molecular Weight: 150,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016546
CAS-Nummer: 24634-61-5
https://de.vwr.com/stibo/web/std.lang.all/71/23/2297123.jpg

Bestellen

Spezifikation Testergebnisse

Assay (Dried Basis) 98.0 - 101.0%
Acidity To Pass Test
Alkalinity To Pass Test
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR