Pressure filter holders with reservoir, stainless steel
Lieferant: Advantec
ADVT302300EA 0 EUR
ADVT302300
Pressure filter holders with reservoir, stainless steel
Filterhalter
These quality stainless steel holders are designed for filtering small batches of liquid without having to use a separate pressure vessel.
  • Suitable for hazardous toxicity testing and TCLP
  • KST142 screen is PTFE coated
  • Use for sterilising, ultracleaning, and flow decay testing for pilot studies
There are separate ports for pressurizing the reservoir and for adding additional sample. A standard silicone O-ring seals the membrane internally by hand-tightening locking wing nuts. When using for intermittent fill operations, a back pressure support is recommended. Available in 304 stainless steel.