Fluorescent Light, Solid-State, Colibri 7
Lieferant: Zeiss
ZEIS4230529741000EA 0 EUR
ZEIS4230529741000
Fluorescent Light, Solid-State, Colibri 7
Beleuchtung Leuchtstofflampen
This solid-state light source Colibri 7 is designed with integrated control unit for continuous brightness adjustment, quickly switchable and stable on the long term.
  • Integrated monitor diode for repetitious brightness adjustment
  • Equipped with 7 excitation lines
Red (630 nm) for excitation of Cy5, Alexa 631, TOTO-3 and similar dyes, yellow (590 nm) for excitation of mCherry, Alexa 568, mPlum and similar dyes, green (555 nm) for excitation of Cy3, TRITC, DsRed and similar dyes, cyan (511 nm) for excitation of eYFP, Eosin, TOTO-1 and similar dyes, blue (475 nm) for excitation of eGFP, Fluo4, FITC and similar dyes, violet (430 nm) for excitation of eCFP, lucifer yellow, Alexa 430 and similar dyes, UV (385 nm) for excitation of DAPI, Alexa 405, Hoechst 33258 and similar dyes.