BlueShadow High-Pressure Dosing Pump with 10 ml Pump Head
Lieferant: Knauer
KNAUAPC40EIEA 11720 EUR
KNAUAPC40EI
BlueShadow High-Pressure Dosing Pump with 10 ml Pump Head
Pumpen Fass/Transferpumpen
The BlueShadow Pump 40P was designed to provide exceptionally precise and reliable solvent delivery for a wide range of HPLC and dosing applications.
  • Pump head made of stainless steel or with ceramic inlays
  • Liquid transport with low pulsation, stable flow rate and high flow accuracy
  • High physical and chemical stability
  • Direct pump control via analog and digital signals
  • Automatic piston backflushing
Exchangeable pump heads with maximum flow rates of 10 and 50 ml/min cover a wide range of high-pressure dosing tasks. Practically pulse-free flow is achieved by our enhanced smart drive control which actively prevents pressure ripple instead of just dampening it. The improved design ensures a low dead volume and the inline filter protects your column from clogging and pressure build-up.
Pump heads for high-temperature applications enable solivent delivery at up to 120 °C. Minimize temperature gradients by adding a pump head heater and insulation sleeve.