Furaptra (Mag-Fura-2) tetrapotassium salt magnesium indicator
Lieferant: AAT BIOQUEST INC
Achtung
AATB20380EA 295 EUR
AATB20380
Furaptra (Mag-Fura-2) tetrapotassium salt magnesium indicator
Furaptra (Mag-Fura-2) tetrapotassium salt
Mag-Fura-2, tetrapotassium salt is an intracellular magnesium indicator that is ratiometric and UV light-excitable.
  • Cell-permeable
  • Useful for both magnesium and high-concentration calcium detection
Formel: C₁₈H₁₀K₄N₂O₁₁
Molecular Weight: 587 g/mol
CAS-Nummer: 132319-57-4
ADR: 5,II
https://de.vwr.com/stibo/web/std.lang.all/32/11/43933211.jpg