Sulphorhodamine B acid chloride ≥95%

Lieferant: AAT BIOQUEST INC
Achtung

AATB470EA 146 EUR
AATB470
Sulphorhodamine B acid chloride ≥95%
Sulphorhodamine B acid chloride

SRB sulfonyl chloride (also called Lissamine rhodamine sulfonyl chloride™) is highly fluorescent.

Formel: C₂₇H₂₉ClN₂O₆S₂
Molecular Weight: 577,107 g/mol
CAS-Nummer: 62796-29-6
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR