Rhodamin 6G ≥90%
Lieferant: AAT BIOQUEST INC
|
Achtung
|
Synonyme:
9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthyliumchlorid
AATB70EA
115
EUR
AATB70
Rhodamin 6G ≥90%
Rhodamin 6G
Rhodamine 6G is also called Rhodamine 590, R6G, Basic Rhodamine Yellow, or C.I.
Formel:
C₂₈H₃₁ClN₂O₃ Molecular Weight: 479,02 g/mol |
MDL:
MFCD00012665 CAS-Nummer: 989-38-8 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...