CytoTrace™ Red CFDA
Lieferant: AAT Bioquest
AATB22016EA 295 EUR
AATB22016
CytoTrace™ Red CFDA
Tests Zelltests
CytoTrace™ Red fluorescent probe retains the spectral properties of Cy3/TRITC, thus its signal can be easily monitored using fluorescence microscopy or flow cytometry.
  • High Multigenerational Cell Retention
  • Bright Signal
Bitte beachten For research use only.