Spexyte™ Intracellular pH Calibration Buffer Kit
Lieferant: AAT Bioquest
AATB21235EA 295 EUR
AATB21235
Spexyte™ Intracellular pH Calibration Buffer Kit
Tests Zelltests
Intracellular pH (pHi) plays an important modulating role in many cellular events, including cell volume regulation, cellular metabolism, calcium regulation, receptor-mediated signal transduction, ion transport, endocytosis, and other cellular processes.
  • Suitable for all major pH indicators
  • Sensitive and quick results
  • Includes a range of calibration buffers
Bitte beachten For research use only.