Buccutite™ FOL-Dye 650 (2 µmoles)
Lieferant: AAT Bioquest
Buccutite™
AATB5372EA 295 EUR
AATB5372
Buccutite™ FOL-Dye 650 (2 µmoles)
Proteinkonjugate und Markierungen
Buccutite™crosslinking technology provides the most convenient and effective crosslinking method to link two biomolecules with a high conjugation yield.
  • High conjugation yield
  • Stable crosslinking
  • Minimal hands-on time
  • Robust protocol