Stains-all ≥95%

Lieferant: AAT BIOQUEST INC
Achtung

AATB10050EA 115 EUR
AATB10050
Stains-all ≥95%
Stains-all

Stains-all is suitable for differential staining of nucleic acids and proteins.

Formel: C₃₀H₂₇BrN₂S₂
Molecular Weight: 559,584 g/mol
MDL: MFCD00078788
CAS-Nummer: 7423-31-6
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR