Bimsstein, BAKER, Laborreagens, J.T.Baker®
Lieferant: Avantor
Synonyme:
Bims
1926.1000EA
114.87
EUR
1926.1000
Bimsstein, BAKER, Laborreagens, J.T.Baker®
Pumice stone
Schmelzpunkt:
> 500 °C |
MDL:
MFCD00148147 CAS-Nummer: 1332-09-8 |
Spezifikation Testergebnisse
Acid-soluble substances | max. 6.0 % |
Iron (Fe) | passes test |
Water Soluble Substances | max. 0.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...