Sulphorhodamine 101 ≥95%
Lieferant: AAT BIOQUEST INC
AATB77EA
109
EUR
AATB77
Sulphorhodamine 101 ≥95%
Sulphorhodamine 101
Sulforhodamine 101 is a fluorescence reference standard for measuring fluorescence quantum yield.
- Fluorescence Reference Standard
Bitte beachten For research use only.
Formel:
C₃₁H₃₀N₂O₇S₂ Molecular Weight: 606,708 g/mol |
MDL:
MFCD00012407 CAS-Nummer: 60311-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...