Pneumatic Valves Tube, CR8
Lieferant: Entegris
ENTECR8-3CT-8FFFEA 0 EUR
ENTECR8-3CT-8FFF
Pneumatic Valves Tube, CR8
Ventile Elliptische Ventile
Designed to divert one media stream into two directions, or alternate two media streams into one, minimizing the footprint. Handles temperatures up to 130 °C (266 °F).
  • Compact 3-way design saves valuable space on OEM equipment
  • Sealed actuator with vent port eliminates chemical exposure of the valve spring
  • Modular design enables manifold customization
  • A variety of connections including PrimeLock®, Flaretek® and Super 300 Type Pillar® fitting